Learn More
Medchemexpress LLC HY-P7525 10ug Medchemexpress, Apolipoprotein A-I, His (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP752510UG
Encompass_Preferred
Medchemexpress, HY-P7525 10ug Apolipoprotein A-I, His (HEK293-expressed), Human CAS: Recombinant Human Apolipoprotein A-I, His (HEK293-expressed) expresses in HEK293 with a His tag at the N-terminus. Apolipoprotein A-I (ApoA-I), the major protein component in high-density lipoprotein (HDL), plays key roles in the Reverse Cholesterol Transport pathway. Apolipoprotein A-I is associated with dengue virus (DV) particles and enhances virus infection through SR-BI. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3173