Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein A-I (mouse) | 20 UG
SDP

Catalog No. 5000291425
Encompass_Preferred

Recombinant mouse apolipoprotein A-I (APOA1), His-tagged and expressed in HEK293 cells, is supplied as a sterile, filtered solution for research applications involving lipid metabolism, cholesterol transport, and protein interaction studies.

  • Recombinant mouse apolipoprotein A-I expressed in HEK293 cells.
  • His tag for purification and detection.
  • Supplied as a 0.22 μm filtered PBS solution with 5% trehalose, 5% mannitol, and 0.01% Tween-80.
  • Endotoxin level <1 EU/μg as determined by LAL.
  • Store at -80 °C; shipped on dry ice; avoid repeated freeze-thaw cycles.
  • Intended for research use in cholesterol efflux and HDL function assays.
  • Available in a 20 μg vial.

Catalog No. 50-002-91425 Supplier Medchemexpress LLC Supplier No. HYP7550320UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant mouse apolipoprotein A-I (APOA1), His-tagged and expressed in HEK293 cells, is supplied as a sterile, filtered solution for research applications involving lipid metabolism, cholesterol transport, and protein interaction studies.

  • Recombinant mouse apolipoprotein A-I expressed in HEK293 cells.
  • His tag for purification and detection.
  • Supplied as a 0.22 μm filtered PBS solution with 5% trehalose, 5% mannitol, and 0.01% Tween-80.
  • Endotoxin level <1 EU/μg as determined by LAL.
  • Store at -80 °C; shipped on dry ice; avoid repeated freeze-thaw cycles.
  • Intended for research use in cholesterol efflux and HDL function assays.
  • Available in a 20 μg vial.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.