Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Apolipoprotein A-II, Human Plasma, 95%

Catalog No. AAJ64658MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64658MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64658MCR Supplier Thermo Scientific™ Supplier No. J64658MCR

missing translation for 'validMainframeMsg'

For research use

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
Formulation Aqueous solution
Quantity 1 mg
Source Human plasma
Storage Requirements -30° to -10°C
Regulatory Status RUO
Product Type Apolipoprotein A-II, Human Plasma, 95%
Recombinant Native
Form Liquid
Purity or Quality Grade 95%

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.