Learn More
Cusabio Technology LLC apolipoprotein A1 Apo-A1
Catalog No. 5000051336
Encompass_Preferred
This kit detects Apo-A1 in monkey serum and plasma samples using a sandwich ELISA method offering a quantitative detection range of 0 7 ng/mL to 500 ng/mL with a sensitivity of 0 7 ng/mL Designed for efficient analysis the assay requires a sample volume of 50-100 L and completes within 1-5 hours of incubation time Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable and precise results for research applications involving apolipoprotein A1 quantification in primate biological samples
Catalog No. 50-000-51336 Supplier Cusabio Technology LLC Supplier No. CSBE11041MO
This kit detects Apo-A1 in monkey serum and plasma samples using a sandwich ELISA method offering a quantitative detection range of 0 7 ng/mL to 500 ng/mL with a sensitivity of 0 7 ng/mL Designed for efficient analysis the assay requires a sample volume of 50-100 L and completes within 1-5 hours of incubation time Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable and precise results for research applications involving apolipoprotein A1 quantification in primate biological samples