Learn More
ABclonal Technology Apolipoprotein A2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A146901000UL
Encompass_Preferred
This gene encodes apolipoprotein (apo-) A-II, which is the second most abundant protein of the high density lipoprotein particles. The protein is found in plasma as a monomer, homodimer, or heterodimer with apolipoprotein D. Defects in this gene may result in apolipoprotein A-II deficiency or hypercholesterolemia.
Catalog No. 50-001-07652