Learn More
Biomatik Corporation Apolipoprotein A4 (Apoa4) Monoclonal Antibody
Supplier:  Biomatik Corporation CAU29389
Encompass_Preferred
Apolipoprotein A4 (Apoa4) Monoclonal Antibody is a Pig (Sus scrofa; Porcine) Monoclonal Antibody to Apolipoprotein A4 (APOA4). The Apolipoprotein A4 (Apoa4) Monoclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Mouse. Species: Pig (Sus scrofa; Porcine). Immunogen: Glu21~Asn235. Applications: WB, IHC, ICC, IP. Target Synonyms: Apolipoprotein A4 (APOA4). Accession Number: O46409. Gene ID: 397681. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9215