Learn More
CPC Scientific H-Lys-Tyr-Tyr-Glu-Leu-Glu-Glu-Lys-Ile-Val-Ser-Leu-Ile-Lys-Asn-Leu-Leu-Val-Ala-Leu-Lys-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific APOL001A
Encompass
SEQUENCE: H-Lys-Tyr-Tyr-Glu-Leu-Glu-Glu-Lys-Ile-Val-Ser-Leu-Ile-Lys-Asn-Leu-Leu-Val-Ala-Leu-Lys-OH (trifluoroacetate salt)ONE-LETTER SEQUENCE: KYYELEEKIVSLIKNLLVALKMOLECULAR FORMULA: C119H200N26O32MOLECULAR WEIGHT: 2507STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: SYNONYMS: RESEARCH AREA: Alzheimer's DiseaseREFERENCES:SKU(s): APOL-001A, APOL-001B
Catalog No. 50-210-8784