Learn More
Cusabio Technology LLC apolipoprotein B100
Catalog No. 5000049510
Encompass_Preferred
This ELISA kit is designed for the quantitative detection of Rat apolipoprotein B100 (Apo-B100) in serum plasma and tissue homogenates using a competitive assay format The kit demonstrates a detection range of 0 313 g/mL to 20 g/mL with a sensitivity threshold of 0 342 g/mL enabling precise measurement of low-abundance samples The protocol requires 50-100 L of sample per test and delivers results within 1-5 hours through absorbance readings at 450 nm Its optimized competitive binding methodology ensures specific identification of APOB while minimizing cross-reactivity making it suitable for research applications involving lipid metabolism studies cardiovascular disease models and biomarker validation in rodent systems
Catalog No. 50-000-49510 Supplier Cusabio Technology LLC Supplier No. CSBE08102R
This ELISA kit is designed for the quantitative detection of Rat apolipoprotein B100 (Apo-B100) in serum plasma and tissue homogenates using a competitive assay format The kit demonstrates a detection range of 0 313 g/mL to 20 g/mL with a sensitivity threshold of 0 342 g/mL enabling precise measurement of low-abundance samples The protocol requires 50-100 L of sample per test and delivers results within 1-5 hours through absorbance readings at 450 nm Its optimized competitive binding methodology ensures specific identification of APOB while minimizing cross-reactivity making it suitable for research applications involving lipid metabolism studies cardiovascular disease models and biomarker validation in rodent systems