Learn More
Biomatik Corporation Apolipoprotein B100 (Apob100) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU29063
Encompass_Preferred
Apolipoprotein B100 (Apob100) Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Apolipoprotein B100 (APOB100). The Apolipoprotein B100 (Apob100) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: His3365~Glu3548. Applications: WB, IHC, ICC, IP. Target Synonyms: Apolipoprotein B100 (APOB100). Accession Number: P04114. Gene ID: 338. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7079