Learn More
Cusabio Technology LLC apolipoprotein C-II
Catalog No. 5000051702
Encompass_Preferred
This kit detects APOC2 in mouse serum plasma and tissue homogenates with a detection range of 18 8 ng/mL to 1200 ng/mL offering a sensitivity of 9 4 ng/mL for precise quantitative analysis Utilizing a competitive ELISA format the assay requires 50-100 L of sample per test and achieves results within 1-5 hours of incubation time The detection is performed at 450 nm wavelength ensuring reliable measurement of apolipoprotein C-II concentrations in biological samples Designed for research applications it provides a standardized method to assess APOC2 levels supporting studies related to lipid metabolism and cardiovascular health The kit s optimized protocol balances speed and accuracy making it suitable for laboratories requiring efficient quantification of this biomarker
Catalog No. 50-000-51702 Supplier Cusabio Technology LLC Supplier No. CSBEL001932MO
This kit detects APOC2 in mouse serum plasma and tissue homogenates with a detection range of 18 8 ng/mL to 1200 ng/mL offering a sensitivity of 9 4 ng/mL for precise quantitative analysis Utilizing a competitive ELISA format the assay requires 50-100 L of sample per test and achieves results within 1-5 hours of incubation time The detection is performed at 450 nm wavelength ensuring reliable measurement of apolipoprotein C-II concentrations in biological samples Designed for research applications it provides a standardized method to assess APOC2 levels supporting studies related to lipid metabolism and cardiovascular health The kit s optimized protocol balances speed and accuracy making it suitable for laboratories requiring efficient quantification of this biomarker