Learn More
Medchemexpress LLC HY-P7529 10ug Medchemexpress, Apolipoprotein C-II, His, Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP752910UG
Encompass_Preferred
Medchemexpress, HY-P7529 10ug Apolipoprotein C-II, His, Human CAS: Recombinant Human Apolipoprotein C-II, His is expressed in E. coli with a His tag at the N-terminus. Recombinant Human Apolipoprotein C-II has a pharmacological application for the treatment of patients with genetic hypertriglyceridemia caused by ApoC-II deficiency. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3174