Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein C-III 50ug
SDP

Catalog No. 5000195932
Encompass_Preferred

Recombinant mouse apolipoprotein C-III (APOC3) fragment (residues E21-S99) expressed with an N-terminal 6xHis-SUMO tag. Supplied as a purified protein for biochemical and immunological assays, the product is provided in small research sizes with accompanying datasheet, certificate of analysis, and safety documentation.

  • Recombinant fragment corresponding to UniProt P33622, residues E21-S99.
  • N-terminal 6xHis and SUMO tags for purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~22 kDa.
  • Available in small research sizes, including a 50 μg vial.

Catalog No. 50-001-95932 Supplier Medchemexpress LLC Supplier No. HYP7208250UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse apolipoprotein C-III (APOC3) fragment (residues E21-S99) expressed with an N-terminal 6xHis-SUMO tag. Supplied as a purified protein for biochemical and immunological assays, the product is provided in small research sizes with accompanying datasheet, certificate of analysis, and safety documentation.

  • Recombinant fragment corresponding to UniProt P33622, residues E21-S99.
  • N-terminal 6xHis and SUMO tags for purification and detection.
  • Purity greater than 90% as determined by reducing SDS-PAGE.
  • Approximate molecular weight ~22 kDa.
  • Available in small research sizes, including a 50 μg vial.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.