Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Apolipoprotein C3 ELISA Kit

Catalog No. EHAPOC3X5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHAPOC3X5 5 x 96 Tests
EHAPOC3 96 Tests
EHAPOC3X10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EHAPOC3X5 Supplier Invitrogen™ Supplier No. EHAPOC3X5
Add to Cart
Add to Cart

ELISA

The Human Apolipoprotein C3 (Hu ApoC3) ELISA quantitates Hu ApoC3 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ApoC3. Principle of the method The Human ApoC3 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Apolipoprotein C-III, also known as apo-CIII, is a protein that in humans is encoded by the APOC3 gene. Apo-CIII is a component of very low density lipoprotein (VLDL). APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02656
Assay Range 2.44-10000 pg/mL
Assay Sensitivity 2.5 pg/mL
Calibration Range 2.5 pg/mL to 10,000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family GPCR (G-Protein Coupled Receptors)
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human Alpha-2 Microglobulin, ApoA, ApoC1, ApoC2, ApoE, ApoH, Artemin, BMP-6, Calbindin, CK beta 8-1 (CCL23), CNTF, EMMPRIN (CD147), Endostatin, Fibrinogen, FLRG, TNFSF18 (GITR Ligand), GPBB, Hemoglobin, HTRA2, IL-17 E, IL-1 R3 (IL1 R Acp), Lactoferrin, Mesothelin, PAPP-A, PD-ECGF (Thymidine Phosphorylase), PDGF R alpha, Pepsinogen 2, Periostin, Persephin, PTH, RAP, SAA (Serum Amyloid A), Serpin F1 (PEDF), Tie-1, TPA, Thrombospondin 2, Ubiquitin+1, Vitronectin, VWF
Sufficient For 5 x 96 Assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Apoc3; APOCIII; Apo-CIII; apoC-III; Apolipoprotein; Apolipoprotein C3; apolipoprotein C-3; apolipoprotein C-III; HALP2; MGC150353Apoc3; APOCIII; Apo-CIII; apoC-III; Apolipoprotein; Apolipoprotein C3; apolipoprotein C-3; apolipoprotein C-III; HALP2; MGC150353; APOCIII; HALP2
Gene ID (Entrez) 345
Gene Symbol APOC3
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Protein Aliases Apo-CIII, ApoC-III, Apolipoprotein C3
Quantity 5 x 96 Tests
Regulatory Status RUO
Research Discipline Cardiovascular Research
Sample Volume Plasma 2 μL, Serum 2 μL, Supernatant 100 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym APOCIII, HALP2
Target ApoC3
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.