Learn More
Randox Apolipoprotein Calibrator 2 | 3x1ml
SDP
Catalog No. NC1827904
Encompass_Preferred
A multi-analyte calibrator covering four specialised apolipoprotein assays. A reconstituted stability of up to 28 days helps to keep waste and costs to a minimum.
This item is not returnable. View return policy
Catalog No. NC1827904 Supplier Randox Supplier No. LP5047
A multi-analyte calibrator covering four specialised apolipoprotein assays. A reconstituted stability of up to 28 days helps to keep waste and costs to a minimum.