Learn More
Randox Apolipoprotein C-III assay
SDP
Catalog No. NC1827903
Encompass_Preferred
For the quantitative in vitro determination of Apo C-III in serum or plasma. This product is suitable for automated, semi-automated and manual use.
This item is not returnable. View return policy
Catalog No. NC1827903 Supplier Randox Supplier No. LP3865
For the quantitative in vitro determination of Apo C-III in serum or plasma. This product is suitable for automated, semi-automated and manual use.