Learn More
Medchemexpress LLC Apolipoprotein E/APO 100ug | 100ug
SDP
Catalog No. 5000187730
Encompass_Preferred
Apolipoprotein E (APOE) plays a key role in lipoprotein-mediated lipid transport and is a core component in the production transformation and clearance of plasma lipoproteins As an amphipathic molecule APOE binds to various lipoprotein particles including chylomicrons chylomicron remnants VLDL and IDL favoring HDL Apolipoprotein E/APOE Protein Human (Biotinylated HEK293 His) is the recombinant human-derived Apolipoprotein E/APOE protein expressed by HEK293 with N-His labeled tag
Catalog No. 50-001-87730 Supplier Medchemexpress LLC Supplier No. HYP78062100UG
Apolipoprotein E (APOE) plays a key role in lipoprotein-mediated lipid transport and is a core component in the production transformation and clearance of plasma lipoproteins As an amphipathic molecule APOE binds to various lipoprotein particles including chylomicrons chylomicron remnants VLDL and IDL favoring HDL Apolipoprotein E/APOE Protein Human (Biotinylated HEK293 His) is the recombinant human-derived Apolipoprotein E/APOE protein expressed by HEK293 with N-His labeled tag