Learn More
Medchemexpress LLC Apolipoprotein E, rat, His-tagged | >90.0% | 20UG
SDP
Catalog No. 5000176400
Encompass_Preferred
Recombinant rat apolipoprotein E (His-tagged) supplied lyophilized for research applications. The protein is expressed in E. coli, has an approximate molecular weight of 45 kDa, and is purified to greater than 90% by reducing SDS-PAGE. The product is lyophilized from a buffered solution containing trehalose and is intended for biochemical and lipid metabolism studies; follow recommended reconstitution and storage conditions.
Catalog No. 50-001-76400 Supplier Medchemexpress LLC Supplier No. HYP70109620UG
Recombinant rat apolipoprotein E (His-tagged) supplied lyophilized for research applications. The protein is expressed in E. coli, has an approximate molecular weight of 45 kDa, and is purified to greater than 90% by reducing SDS-PAGE. The product is lyophilized from a buffered solution containing trehalose and is intended for biochemical and lipid metabolism studies; follow recommended reconstitution and storage conditions.