Learn More
Medchemexpress LLC Apolipoprotein E/APOE4 protein, human (HEK293, His) | >95.0% | 20UG
SDP
Catalog No. 5000174941
Encompass_Preferred
Recombinant human apolipoprotein E (APOE4) produced in HEK293 cells, featuring an N-terminal polyhistidine tag. Supplied as a lyophilized powder for research use, it offers high purity and native-like post-translational modifications suitable for biochemical and cell-based assays.
  • Supplied as a lyophilized powder for convenient storage.
  • Produced in human HEK293 cells to preserve native post-translational modifications.
  • N-terminal His tag enables affinity purification and detection.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~35-40 kDa due to glycosylation.
  • Suitable for biochemical assays and structural studies.
  • Store frozen and aliquot to minimize freeze-thaw cycles.
Catalog No. 50-001-74941 Supplier Medchemexpress LLC Supplier No. HYP70085320UG
Recombinant human apolipoprotein E (APOE4) produced in HEK293 cells, featuring an N-terminal polyhistidine tag. Supplied as a lyophilized powder for research use, it offers high purity and native-like post-translational modifications suitable for biochemical and cell-based assays.
  • Supplied as a lyophilized powder for convenient storage.
  • Produced in human HEK293 cells to preserve native post-translational modifications.
  • N-terminal His tag enables affinity purification and detection.
  • Purity >95.0% as determined by reducing SDS-PAGE.
  • Apparent molecular weight ~35-40 kDa due to glycosylation.
  • Suitable for biochemical assays and structural studies.
  • Store frozen and aliquot to minimize freeze-thaw cycles.