Learn More
Medchemexpress LLC Apolipoprotein E (APOE2) recombinant human protein (R154S, R176C), C-terminal His-tag | >90.0% | 50UG
SDP
Supplier:  Medchemexpress LLC HYP70109450UG
Encompass_Preferred
Recombinant human apolipoprotein E (APOE2 variant R154S, R176C) is supplied as a C-terminal His-tagged protein expressed in HEK293 cells. The product is lyophilized from PBS-based formulations, displays mammalian glycosylation (≈33-42 kDa), and is intended for research on lipid metabolism and APOE isoform biology. Purity is >90% by reducing SDS-PAGE, and multiple package sizes are available.
Catalog No. 50-001-75330