Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein E/APOE3 protein, human (HEK293, hFc) | >95.0% | 50UG
SDP

Catalog No. 5000176761
Encompass_Preferred

Recombinant human apolipoprotein E (APOE3) with an N-terminal human Fc tag, expressed in HEK293 cells and supplied lyophilized. The protein is formulated in PBS with 8% trehalose, has >95% purity, and a reported molecular weight of 60-70 kDa. It is intended for research use and has demonstrated binding to human TREM2 in SPR assays.

  • Recombinant human APOE3 with N-terminal hFc tag.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Lyophilized from PBS with 8% trehalose for improved stability.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Demonstrated binding to human TREM2 in SPR assays.
  • Stable when stored frozen; aliquot and store at -20°C or -80°C for long-term storage.

Catalog No. 50-001-76761 Supplier Medchemexpress LLC Supplier No. HYP70105150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human apolipoprotein E (APOE3) with an N-terminal human Fc tag, expressed in HEK293 cells and supplied lyophilized. The protein is formulated in PBS with 8% trehalose, has >95% purity, and a reported molecular weight of 60-70 kDa. It is intended for research use and has demonstrated binding to human TREM2 in SPR assays.

  • Recombinant human APOE3 with N-terminal hFc tag.
  • Expressed in HEK293 cells for human-like post-translational modifications.
  • Lyophilized from PBS with 8% trehalose for improved stability.
  • Purity >95% as determined by reducing SDS-PAGE.
  • Demonstrated binding to human TREM2 in SPR assays.
  • Stable when stored frozen; aliquot and store at -20°C or -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.