Recombinant rabbit apolipoprotein E (APOE) is supplied as a lyophilized, affinity- and epitope-tagged protein for biochemical and cell biology research focused on lipid transport, receptor binding, and APOE-related assays. The product is expressed in E. coli and provided with formulation and storage guidance to support reproducible experimental use.
- Recombinant rabbit apolipoprotein E suitable for lipid transport and receptor-binding studies.
- Expressed in E. coli for high-yield production.
- Includes affinity and epitope tags for purification and detection.
- Full-length mature sequence (T20-Q311) representing native protein regions.
- Approximate molecular weight ~55 kDa.
- Purity greater than 90% by reducing SDS-PAGE.
- Lyophilized formulation with stabilizers for room-temperature handling prior to reconstitution.
- Stable storage recommended at -20°C; aliquot and freeze at -20°C or -80°C for extended storage.