Learn More
Medchemexpress LLC Apolipoprotein E (APOE3), human recombinant | 95.0% | 100 UG
SDP
Catalog No. 5000291055
Encompass_Preferred
Recombinant human apolipoprotein E (APOE3) produced in HEK293 cells with an N-terminal His tag. Supplied as a purified protein for research use in studies of lipid metabolism, neurobiology, and protein interactions. Molecular weight ~36-39 kDa and listed purity 95%.
Catalog No. 50-002-91055 Supplier Medchemexpress LLC Supplier No. HYP7531100UG
Recombinant human apolipoprotein E (APOE3) produced in HEK293 cells with an N-terminal His tag. Supplied as a purified protein for research use in studies of lipid metabolism, neurobiology, and protein interactions. Molecular weight ~36-39 kDa and listed purity 95%.