Learn More
Medchemexpress LLC Apolipoprotein E/APOE3 protein, human (HEK293, His) | ≥ 95% | Approximately 36-40 kDa | 500 UG
SDP
Catalog No. 5000399883
Encompass_Preferred
The product is a recombinant human Apolipoprotein E (ApoE3) protein, expressed in HEK293 cells with a His tag. It plays a vital role in lipid transport and homeostasis in both plasma and the central nervous system by interacting with various LDL receptors and heparin. This protein is essential for cholesterol clearance and immune response modulation.
  • It is a recombinant protein with an N-terminal His tag.
  • It mediates lipid transport and homeostasis.
  • It binds to LDL receptors, LRP1, LRP2, LRP8, VLDLR, and heparin.
  • It promotes HDL synthesis and cholesterol clearance.
  • It can modulate immune responses.
  • It is suitable for research use.
  • Has a purity of ≥ 95%.
  • Has an endotoxin level of <1 EU/μg.
  • Stable at -20°C for 2 years.
  • Biological activity measured in SH-SY5Y cells with an ED50 of 15.14 ng/mL.
Catalog No. 50-003-99883 Supplier Medchemexpress LLC Supplier No. HYP7531500UG
The product is a recombinant human Apolipoprotein E (ApoE3) protein, expressed in HEK293 cells with a His tag. It plays a vital role in lipid transport and homeostasis in both plasma and the central nervous system by interacting with various LDL receptors and heparin. This protein is essential for cholesterol clearance and immune response modulation.
  • It is a recombinant protein with an N-terminal His tag.
  • It mediates lipid transport and homeostasis.
  • It binds to LDL receptors, LRP1, LRP2, LRP8, VLDLR, and heparin.
  • It promotes HDL synthesis and cholesterol clearance.
  • It can modulate immune responses.
  • It is suitable for research use.
  • Has a purity of ≥ 95%.
  • Has an endotoxin level of <1 EU/μg.
  • Stable at -20°C for 2 years.
  • Biological activity measured in SH-SY5Y cells with an ED50 of 15.14 ng/mL.