Learn More
Medchemexpress LLC Apolipoprotein E (mouse) | >90.0% | 100 UG
SDP
Catalog No. 5000291147
Encompass_Preferred
Recombinant mouse apolipoprotein E (APOE), C-terminal His-tagged and expressed in HEK293 cells. Supplied lyophilized with trehalose and formulated in buffered saline, this protein is intended for biochemical assays, binding studies, and cell-based activity measurements. Purity is >90% by reducing SDS-PAGE, with an estimated molecular weight of ~34-40 kDa. Store at -20°C; aliquot and freeze for extended storage.
Catalog No. 50-002-91147 Supplier Medchemexpress LLC Supplier No. HYP7532100UG
Recombinant mouse apolipoprotein E (APOE), C-terminal His-tagged and expressed in HEK293 cells. Supplied lyophilized with trehalose and formulated in buffered saline, this protein is intended for biochemical assays, binding studies, and cell-based activity measurements. Purity is >90% by reducing SDS-PAGE, with an estimated molecular weight of ~34-40 kDa. Store at -20°C; aliquot and freeze for extended storage.