Learn More
Sigma Aldrich Fine Chemicals Biosciences Apolipoprotein E4 human re
Catalog No. 502904966
Encompass
Apolipoprotein E (ApoE) is present in the brain and is mainly produced by astrocytes. It is a 299 amino acid glycoprotein of 34kDa. It is present in all classes of lipoproteins except LDL (low-density lipoprotein). APOEnbspgene has three alleles such as APOEnbspε3 APOEnbspε4nbspand APOEnbspε2. It is located on human chromosome 19q13.
Catalog No. 50-290-4966 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. A3234100UG
Apolipoprotein E (ApoE) is present in the brain and is mainly produced by astrocytes. It is a 299 amino acid glycoprotein of 34kDa. It is present in all classes of lipoproteins except LDL (low-density lipoprotein). APOEnbspgene has three alleles such as APOEnbspε3 APOEnbspε4nbspand APOEnbspε2. It is located on human chromosome 19q13.