Learn More
Cusabio Technology LLC apolipoprotein H Apo-H
Catalog No. 5000049432
Encompass_Preferred
The Mouse apolipoprotein H (Apo-H) ELISA Kit is designed for the quantitative measurement of APOH in serum plasma and tissue homogenates using a sandwich ELISA technique This kit detects APOH within a broad detection range of 15 6 ng/mL to 1000 ng/mL with a sensitivity of 3 9 ng/mL ensuring precise and reliable results for low-abundance samples The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours making it suitable for high-throughput applications Measurements are performed at 450 nm and the kit includes all necessary components for efficient APOH quantification Ideal for research in lipid metabolism cardiovascular diseases or autoimmune disorders this kit provides a robust tool for studying APOH dynamics in diverse biological matrices
Catalog No. 50-000-49432 Supplier Cusabio Technology LLC Supplier No. CSBE10074M
The Mouse apolipoprotein H (Apo-H) ELISA Kit is designed for the quantitative measurement of APOH in serum plasma and tissue homogenates using a sandwich ELISA technique This kit detects APOH within a broad detection range of 15 6 ng/mL to 1000 ng/mL with a sensitivity of 3 9 ng/mL ensuring precise and reliable results for low-abundance samples The assay requires a sample volume of 50-100 L and delivers results within 1-5 hours making it suitable for high-throughput applications Measurements are performed at 450 nm and the kit includes all necessary components for efficient APOH quantification Ideal for research in lipid metabolism cardiovascular diseases or autoimmune disorders this kit provides a robust tool for studying APOH dynamics in diverse biological matrices