Learn More
Sino Biological Apolipoprotein H / APOH Antibody, Rabbit MAb 100uL
Supplier:  Sino Biological 11221R003100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Apolipoprotein H / APOH (rh Apolipoprotein H / APOH; Catalog#11221-H08H; NP_000033.2; Met 1-Cys345).
Catalog No. 50-224-8239