Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein H/APOH Protein, Human (HEK293, His) | >= 95% | Approximately 45-70 kDa | 250 UG
SDP

Catalog No. 5000397313
Encompass_Preferred

Apolipoprotein H, also known as APOH, is a human protein expressed in HEK293 cells with a His tag at the C-terminus. This protein is a primary target of anti-phospholipid antibodies (aPLs) associated with systemic lupus erythematosus and antiphospholipid syndrome. It is supplied as a lyophilized powder, formulated from a 0.22 μm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.2; or 50 mM Tris-HCl, 300 mM NaCl, pH 7.4; or PBS, pH 7.4, 8% trehalose.

  • Expressed in HEK293 cells with a C-terminal His tag
  • Purity of >= 95% as determined by reducing SDS-PAGE
  • Molecular weight approximately 45-70 kDa
  • Endotoxin level less than 1 EU/μg
  • Functional in cell proliferation assay with Jurkat human T-lymphocyte leukemia cells
  • Binds Biotinylated Human LDL R in functional ELISA

Catalog No. 50-003-97313 Supplier Medchemexpress LLC Supplier No. 250UGHYP7533
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Apolipoprotein H, also known as APOH, is a human protein expressed in HEK293 cells with a His tag at the C-terminus. This protein is a primary target of anti-phospholipid antibodies (aPLs) associated with systemic lupus erythematosus and antiphospholipid syndrome. It is supplied as a lyophilized powder, formulated from a 0.22 μm filtered solution of 20 mM PB, 150 mM NaCl, pH 7.2; or 50 mM Tris-HCl, 300 mM NaCl, pH 7.4; or PBS, pH 7.4, 8% trehalose.

  • Expressed in HEK293 cells with a C-terminal His tag
  • Purity of >= 95% as determined by reducing SDS-PAGE
  • Molecular weight approximately 45-70 kDa
  • Endotoxin level less than 1 EU/μg
  • Functional in cell proliferation assay with Jurkat human T-lymphocyte leukemia cells
  • Binds Biotinylated Human LDL R in functional ELISA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.