Learn More
Medchemexpress LLC Apolipoprotein M (APOM) | >90.0% | 100 UG
SDP
Catalog No. 5000293016
Encompass_Preferred
Recombinant human apolipoprotein M supplied as a lyophilized, His-tagged protein produced in HEK293 cells. The product represents full-length isoform-1 (residues M1-N188), is >90% pure by reducing SDS-PAGE, and is lyophilized from PBS pH 7.4 for stable storage. Intended for research use in biochemical and structural studies.
  • Recombinant human apolipoprotein M expressed in HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Full-length isoform-1 (residues M1-N188).
  • Greater than 90.0% purity by reducing SDS-PAGE.
  • Lyophilized formulation for enhanced stability.
  • Lyophilized from PBS, pH 7.4 (0.22 μm filtered) for consistent reconstitution.
Catalog No. 50-002-93016 Supplier Medchemexpress LLC Supplier No. HYP7536100UG
Recombinant human apolipoprotein M supplied as a lyophilized, His-tagged protein produced in HEK293 cells. The product represents full-length isoform-1 (residues M1-N188), is >90% pure by reducing SDS-PAGE, and is lyophilized from PBS pH 7.4 for stable storage. Intended for research use in biochemical and structural studies.
  • Recombinant human apolipoprotein M expressed in HEK293 cells.
  • C-terminal 6xHis tag for detection and purification.
  • Full-length isoform-1 (residues M1-N188).
  • Greater than 90.0% purity by reducing SDS-PAGE.
  • Lyophilized formulation for enhanced stability.
  • Lyophilized from PBS, pH 7.4 (0.22 μm filtered) for consistent reconstitution.