Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein M (APOM) protein, human | 97.0% | 5 UG
SDP

Catalog No. 5000291359
Encompass_Preferred

Recombinant human apolipoprotein M (APOM), 166 amino acids, expressed in HEK293 cells and supplied as a lyophilized, His-tagged protein for research applications. The product is characterized for purity and stability and includes handling and storage recommendations.

  • Human APOM sequence (166 amino acids).
  • Expressed in HEK293 mammalian cells for native post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Lyophilized powder format for ease of storage and transport.
  • High purity (~97%) as determined by reducing SDS-PAGE.
  • Recommended storage at -20°C; aliquot and store at -80°C for long-term stability.

Catalog No. 50-002-91359 Supplier Medchemexpress LLC Supplier No. HYP7536A5UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human apolipoprotein M (APOM), 166 amino acids, expressed in HEK293 cells and supplied as a lyophilized, His-tagged protein for research applications. The product is characterized for purity and stability and includes handling and storage recommendations.

  • Human APOM sequence (166 amino acids).
  • Expressed in HEK293 mammalian cells for native post-translational modifications.
  • C-terminal 6xHis tag enables affinity purification and detection.
  • Lyophilized powder format for ease of storage and transport.
  • High purity (~97%) as determined by reducing SDS-PAGE.
  • Recommended storage at -20°C; aliquot and store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.