Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Apolipoprotein M/APOM protein, human | 97.0% | 500 UG
SDP

Catalog No. 5000291825
Encompass_Preferred

Recombinant human apolipoprotein M (APOM) supplied lyophilized; expressed in HEK293 cells and provided with a His tag. The protein sequence corresponds to residues C23-N188 (166 amino acids), with an expected molecular weight of 19-24 kDa and low endotoxin suitable for research applications. Store frozen and avoid repeated freeze-thaw cycles.

  • Recombinant human protein expressed in HEK293 cells.
  • His-tagged for detection and purification.
  • Sequence corresponds to C23-N188 (166 amino acids).
  • Expected molecular weight 19-24 kDa.
  • Endotoxin level <1 EU/μg.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Reconstitution recommended at ≥100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.

Catalog No. 50-002-91825 Supplier Medchemexpress LLC Supplier No. HYP7536A500UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human apolipoprotein M (APOM) supplied lyophilized; expressed in HEK293 cells and provided with a His tag. The protein sequence corresponds to residues C23-N188 (166 amino acids), with an expected molecular weight of 19-24 kDa and low endotoxin suitable for research applications. Store frozen and avoid repeated freeze-thaw cycles.

  • Recombinant human protein expressed in HEK293 cells.
  • His-tagged for detection and purification.
  • Sequence corresponds to C23-N188 (166 amino acids).
  • Expected molecular weight 19-24 kDa.
  • Endotoxin level <1 EU/μg.
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4.
  • Reconstitution recommended at ≥100 μg/mL in ddH2O.
  • Store at -20°C; aliquot and freeze at -20°C or -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.