Learn More
Medchemexpress LLC HY-P7536 10ug Medchemexpress, Apolipoprotein M, His (HEK293-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP753610UG
Encompass_Preferred
Medchemexpress, HY-P7536 10ug Apolipoprotein M, His (HEK293-expressed), Human CAS: Recombinant Human Apolipoprotein M, His (HEK293-expressed) expresses in HEK293 with a His tag at the N-terminus. Apolipoprotein E (apoE) secreted by HEK cells stably expressing apoE3 or apoE4 (HEK-apoE) binds Aβ, and inhibits Aβ-induced neurotoxicity . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3177