Learn More
ACROBiosystems ACROBiosystems Human Apolipoprotein E / APOE Protein, His Tag
Catalog No. 501956487
Encompass_Preferred
Recombinant Protein;100UG;Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession # P02649-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.1 kDa.
Catalog No. 50-195-6487 Supplier ACROBiosystems Supplier No. APEH5246100UG
Recombinant Protein;100UG;Human Apolipoprotein E, His Tag (APE-H5246) is expressed from human 293 cells (HEK293). It contains AA Lys 19 - His 317 (Accession # P02649-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 36.1 kDa.