Learn More
ORBITAL BIOSCIENCES LLC Apollo Ultrafiltration Concentrator, 20 mL Capacity, 150 kDa MWCO, 25/Pack
Catalog No. NC9380552
Encompass
The Apollo 20 mL, 150 kDa ultrafiltration concentrator enables efficient concentration and purification of protein solutions and viral gene vectors. With a unique conical design and 100% air-tested membrane integrity, it achieves high concentration factors while minimizing fouling. This device is ideal for research use only, offering over 90% recovery of retained proteins, making it a superior choice for researchers seeking reliable results in their experiments.
Catalog No. NC9380552 Supplier ORBITAL BIOSCIENCES LLC Supplier No. AP2015010
The Apollo 20 mL, 150 kDa ultrafiltration concentrator enables efficient concentration and purification of protein solutions and viral gene vectors. With a unique conical design and 100% air-tested membrane integrity, it achieves high concentration factors while minimizing fouling. This device is ideal for research use only, offering over 90% recovery of retained proteins, making it a superior choice for researchers seeking reliable results in their experiments.