Learn More
Grainger APOLLO Manual Two-Way Ball Valve: 1/2 in, Stainless Steel, Straight Valve Body, FNPT x FNPT, MPTFE
Catalog No. NC3848245
Encompass_Preferred
Stainless steel ball valves are abrasion-resistant, corrosion-resistant, and suitable for high-pressure and high-temperature applications.
Catalog No. NC3848245 Supplier Grainger Supplier No. 3EEZ5
Stainless steel ball valves are abrasion-resistant, corrosion-resistant, and suitable for high-pressure and high-temperature applications.