Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apomorphine Hydrochloride, Hemihydrate, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18600746 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
5 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-746 1 g Amber Glass Bottle
18-600-747 5 g Amber Glass Bottle
18-600-748 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-600-746 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A11471GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

A1147, 41372-20-7, C17H17NO2·HCl·1/2H2O

Spectrum™ Chemical Apomorphine Hydrochloride, Hemihydrate, USP is a non-selective dopamine agonist used in the treatment of Parkinson's disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 41372-20-7
Molecular Weight (g/mol) 312.8 g/mol

Specifications

Packaging Amber Glass Bottle
CAS Max % 100%
UN Number UN2811
Quantity 1 g
Percent Purity 98.5 to 101.5%
Grade USP
Chemical Name or Material Apomorphine Hydrochloride, Hemihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.