Learn More
Cayman Chemical (−)-Apomorphine (hydrochloride hydrate), Purity: ≥95%, CAS: 58117-94-5, 10 mg
SDP
Catalog No. 502968666
Encompass_Preferred
A non-selective dopamine receptor agonist (pKis = 6.43, 7.08, 7.59, 8.36, and 7.83 for human recombinant D1, D2L, D3, D4, and D5 receptors, respectively); produces biphasic effects on locomotor activity and displays anti-Parkinsonian and neuroprotective actions
Catalog No. 50-296-8666 Supplier Cayman Chemical Supplier No. 1609410 MG
A non-selective dopamine receptor agonist (pKis = 6.43, 7.08, 7.59, 8.36, and 7.83 for human recombinant D1, D2L, D3, D4, and D5 receptors, respectively); produces biphasic effects on locomotor activity and displays anti-Parkinsonian and neuroprotective actions