Learn More
Cayman Chemical β-Apooxytetracycline, Purity: ≥98%, CAS: 18751-99-0, 10 mg
SDP
Catalog No. 502940653
Encompass_Preferred
A potential impurity found in commercial preparations of oxytetracycline; has a relative potency of 0.1 compared with oxytetracycline for growth inhibition of aerobic sludge bacteria, an MIC50 value of 32 mg/L for tetracycline-sensitive strains of Pseudomonas, and MIC50 values of greater than 32 mg/L for tetracycline-sensitive strains of Agrobacterium, Moraxella, and Bacillus, as well as tetracycline-resistant strains of E. coli; toxic to rats, decreasing body weight, disrupting blood cell counts, and inducing hepatocyte necrosis
Catalog No. 50-294-0653 Supplier Cayman Chemical Supplier No. 2709910 MG
A potential impurity found in commercial preparations of oxytetracycline; has a relative potency of 0.1 compared with oxytetracycline for growth inhibition of aerobic sludge bacteria, an MIC50 value of 32 mg/L for tetracycline-sensitive strains of Pseudomonas, and MIC50 values of greater than 32 mg/L for tetracycline-sensitive strains of Agrobacterium, Moraxella, and Bacillus, as well as tetracycline-resistant strains of E. coli; toxic to rats, decreasing body weight, disrupting blood cell counts, and inducing hepatocyte necrosis