Learn More
CPC Scientific Ac-Asp-Glu-Val-Asp-AMC 1MG
SDP
Supplier:  CPC Scientific CASP049A
Encompass
Designed substrate based on the tetrapeptide-AMC motif that has been used successfully with caspase-1, using the poly (ADP ribose) polymerase cleavage site P1-P4 tetrapeptide. ONE-LETTER SEQUENCE: Ac-DEVD-AMCMOLECULAR FORMULA: C30H37N5O13MOLECULAR WEIGHT:675.7STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [169332-61-0 ], RESEARCH AREA: ApoptosisREFERENCES: D.W. Nicholson, Nature, 376 (1995); J. Xiang et al., Proc. Natl. Acad. Sci. USA, 93, 14559 (1996)
Catalog No. 50-210-9006