Learn More
Echelon Biosciences Research Labs Apopain Substrate I / Caspase 3 - 5 mg, CAS#: 189950-66-1
SDP
Supplier:  Echelon Biosciences Research Labs 860205MG
Encompass_Preferred
Apopain Substrate I / Caspase 3 - 5mg , Ac-Asp-Glu-Val-Asp-pNA Category/Application: Enzyme Substrates This item is non returnable
Catalog No. 50-190-8862