Learn More
Enzo Life Sciences Apoptosis Activator 2 (5mg). CAS: 79183-19-0
SDP
Catalog No. 502009112
Encompass_Preferred
Induces apoptosis by promoting the oligomerization of Apaf-1 into the mature, functional apoptosome, triggering caspase-9 activation resulting in activated caspase-3. Shows strong selectivity for cancer cells against normal cell lines. Acts in a similar way like apoptosis inducer PETCM (ALX-420-031). Alternative name: 1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione. Purity: ≥98% (HPLC). Appearance: Orange to red solid. Solubility: Soluble in DMSO (5mg/ml), methanol (5mg/ml) or 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-9112 Supplier Enzo Life Sciences Supplier No. ALX420035M005
Induces apoptosis by promoting the oligomerization of Apaf-1 into the mature, functional apoptosome, triggering caspase-9 activation resulting in activated caspase-3. Shows strong selectivity for cancer cells against normal cell lines. Acts in a similar way like apoptosis inducer PETCM (ALX-420-031). Alternative name: 1-[(3,4-Dichlorophenyl)methyl]-1H-indole-2,3-dione. Purity: ≥98% (HPLC). Appearance: Orange to red solid. Solubility: Soluble in DMSO (5mg/ml), methanol (5mg/ml) or 100% ethanol. Long Term Storage: +4°C.