Learn More
Abcam Apoptosis Activator 2 (AAII), Cytochrome c caspase activator and apoptosis inducer, 50MG
Catalog No. 502706780
Encompass_Preferred
MW 306.1 Da, Purity >99%. Cell-permeable cytochrome c caspase activator. Promoting cytochrome-c dependent oligomerization of Apaf-1 into the mature apoptosome. Induces apoptosis in tumor cells (IC₅₀ = ~4 μM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6780 Supplier Abcam Supplier No. AB14122750MG
MW 306.1 Da, Purity >99%. Cell-permeable cytochrome c caspase activator. Promoting cytochrome-c dependent oligomerization of Apaf-1 into the mature apoptosome. Induces apoptosis in tumor cells (IC₅₀ = ~4 μM).
The product is subject to the following: Abcam Restricted Use Statement