Learn More
Cell Signaling Technology Apoptosis Antibody Sampler Kit, Mouse Preferred, 1 Kit
SDP
Catalog No. 502729494
Encompass
The Apoptosis Antibody Sampler Kit (Mouse Specific) is designed for use with mouse samples and provides an economical means to evaluate the levels of active and inactive caspases. The kit contains primary and secondary antibodies sufficient to perform two Western blot experiments for each antibody target.
  • Provides a comprehensive set of antibodies to study apoptosis and caspase signaling pathways.
  • Includes primary antibodies for detecting caspases, cleaved caspases, and cleaved PARP.
  • Supplied with HRP-linked anti-rabbit and anti-mouse IgG secondary antibodies.
  • Optimized for use in Western blotting applications.
  • Supplied in a storage buffer containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide.
Catalog No. 50-272-9494 Supplier Cell Signaling Technology Supplier No. 9930T
The Apoptosis Antibody Sampler Kit (Mouse Specific) is designed for use with mouse samples and provides an economical means to evaluate the levels of active and inactive caspases. The kit contains primary and secondary antibodies sufficient to perform two Western blot experiments for each antibody target.
  • Provides a comprehensive set of antibodies to study apoptosis and caspase signaling pathways.
  • Includes primary antibodies for detecting caspases, cleaved caspases, and cleaved PARP.
  • Supplied with HRP-linked anti-rabbit and anti-mouse IgG secondary antibodies.
  • Optimized for use in Western blotting applications.
  • Supplied in a storage buffer containing 10 mM sodium HEPES (pH 7.5), 150 mM NaCl, 100 μg/mL BSA, 50% glycerol, and <0.02% sodium azide.