Learn More
Biotium Apoptosis Assays Fluorescent Proteins Nucleotides Other Conjugates Annexin V Conjugates CF450 1/EA
Catalog No. 5000500158
Encompass_Preferred
Annexin V fluorescent conjugates can be used to detect apoptotic cells The human anticoagulant Annexin V is a 35-36 kDa calcium-dependent phospholipid-binding protein with high affinity for phosphatidylserine (PS) In normal viable cells PS is located on the inner leaflet of the cytoplasmic membrane In apoptotic cells however PS is translocated from the inner to the outer leaflet of the plasma membrane where it can be detected by fluorescently labeled Annexin V CF Dye Annexin V Conjugates are provided at 50 ug/mL in buffer with azide as a preservative CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes 405 nm-excitable green fluorescent CF450 dye has excitation/emission at 450/538 nm
Catalog No. 50-005-00158 Supplier Biotium Supplier No. 29083
Annexin V fluorescent conjugates can be used to detect apoptotic cells The human anticoagulant Annexin V is a 35-36 kDa calcium-dependent phospholipid-binding protein with high affinity for phosphatidylserine (PS) In normal viable cells PS is located on the inner leaflet of the cytoplasmic membrane In apoptotic cells however PS is translocated from the inner to the outer leaflet of the plasma membrane where it can be detected by fluorescently labeled Annexin V CF Dye Annexin V Conjugates are provided at 50 ug/mL in buffer with azide as a preservative CF dyes are Biotiums line of next-generation fluorescent dyes with advantages in brightness photostability and conjugate specificity compared to other fluorescent dyes 405 nm-excitable green fluorescent CF450 dye has excitation/emission at 450/538 nm