Learn More
Biotium Apoptosis Assays Fluorescent Proteins Nucleotides Other Conjugates Annexin V Conjugates Unlabeled 1/EA
Catalog No. 5000500159
Encompass_Preferred
This product is recombinant Human Annexin V expressed from E coli It is supplied lyophilized and is compatible with dye conjugation to stain phosphatidylserine on the surface of apoptotic cells Unit size 1 mg
Catalog No. 50-005-00159 Supplier Biotium Supplier No. 29089
This product is recombinant Human Annexin V expressed from E coli It is supplied lyophilized and is compatible with dye conjugation to stain phosphatidylserine on the surface of apoptotic cells Unit size 1 mg