Learn More
Biotium Apoptosis Assays Fluorescent Proteins Nucleotides Other Conjugates Annexin V Conjugates Unlabeled 1/EA
Catalog No. 5000500160
Encompass_Preferred
This product is recombinant Human Annexin V expressed from E coli It is supplied lyophilized and is compatible with dye conjugation to stain phosphatidylserine on the surface of apoptotic cells Unit size 5 mg
Catalog No. 50-005-00160 Supplier Biotium Supplier No. 290895MG
Add to Cart
Add to Cart
This product is recombinant Human Annexin V expressed from E coli It is supplied lyophilized and is compatible with dye conjugation to stain phosphatidylserine on the surface of apoptotic cells Unit size 5 mg