Learn More
Biotium Apoptosis Assays Fluorescent Proteins Nucleotides Other Conjugates Annexin V Conjugates Unlabeled 1/EA
Supplier:  Biotium 29089100UG
Encompass_Preferred
This product is recombinant Human Annexin V expressed from E coli It is supplied lyophilized and is compatible with dye conjugation to stain phosphatidylserine on the surface of apoptotic cells Unit size 100 ug
Catalog No. 50-004-99716