Learn More
Selleck Chemical LLC Apoptosis Compound Library L3300-2X100UL/WELL
SDP
Catalog No. 5000041025
Encompass_Preferred
A unique collection of 1223 small molecules used for apoptosis research targeting Bcl-2 Caspase p53 TNF-alpha Mdm2 survivin etc
Catalog No. 50-000-41025 Supplier Selleck Chemical LLC Supplier No. L33002X100UL/WELL
Add to Cart
Add to Cart
Request a Quote
A unique collection of 1223 small molecules used for apoptosis research targeting Bcl-2 Caspase p53 TNF-alpha Mdm2 survivin etc