Learn More
STEMCELL Technologies Annexin V Apoptosis Detection Kit with 7-AAD, APC, Size: 100 Tests
SDP
Catalog No. 502036690
Encompass
This apoptosis staining and detection kit is used for the combined detection of early-stage cell apoptosis (using Annexin V) and late-stage cell apoptosis or necrosis (using both Annexin V and 7-AAD). It includes Annexin V (Catalog #100-0329 or Catalog #100-0331 or Catalog #100-0333), Annexin V Binding Buffer (Catalog #100-0334), and 7-AAD (Catalog #75001). Annexin V belongs to the Annexin family of intracellular proteins that bind to membrane phospholipids in a calcium-dependent manner. This dye has a high affinity for phosphatidylserine (PS) that is present in the inner leaflet of the plasma membrane. During early-stage cell apoptosis, PS is translocated from the inner to the outer leaflet of the cell membrane, exposing it to the external environment, where it is detected by Annexin V. Visit STEMCELL.com for more information and related products.
Catalog No. 50-203-6690 Supplier STEMCELL Technologies Supplier No. 1000339
This apoptosis staining and detection kit is used for the combined detection of early-stage cell apoptosis (using Annexin V) and late-stage cell apoptosis or necrosis (using both Annexin V and 7-AAD). It includes Annexin V (Catalog #100-0329 or Catalog #100-0331 or Catalog #100-0333), Annexin V Binding Buffer (Catalog #100-0334), and 7-AAD (Catalog #75001). Annexin V belongs to the Annexin family of intracellular proteins that bind to membrane phospholipids in a calcium-dependent manner. This dye has a high affinity for phosphatidylserine (PS) that is present in the inner leaflet of the plasma membrane. During early-stage cell apoptosis, PS is translocated from the inner to the outer leaflet of the cell membrane, exposing it to the external environment, where it is detected by Annexin V. Visit STEMCELL.com for more information and related products.