Learn More
Meso Scale Discovery Apoptosis Whole Cell Lysate
SDP
Catalog No. 5000082290
Encompass
The Mesoscale Apoptosis Whole Cell Lysate Kit (K15102D) is a multiplex assay kit that allows for the simultaneous detection of cleaved PARP cleaved caspase-3 phospho-p53 (S15) and total p53 from cell lysates The kit contains a 96-well plate coated with capture antibodies detection antibodies and buffers needed to run the assay This assay kit enables high-throughput quantitative analysis of apoptotic markers and p53 activation status in cells It is compatible with lysates from human mouse and rat samples The kit provides all necessary reagents to analyze 46 samples in duplicate
Catalog No. 50-000-82290 Supplier Meso Scale Discovery Supplier No. K15102D2
Add to Cart
Add to Cart
Request a Quote
The Mesoscale Apoptosis Whole Cell Lysate Kit (K15102D) is a multiplex assay kit that allows for the simultaneous detection of cleaved PARP cleaved caspase-3 phospho-p53 (S15) and total p53 from cell lysates The kit contains a 96-well plate coated with capture antibodies detection antibodies and buffers needed to run the assay This assay kit enables high-throughput quantitative analysis of apoptotic markers and p53 activation status in cells It is compatible with lysates from human mouse and rat samples The kit provides all necessary reagents to analyze 46 samples in duplicate